Would you like to log in again?
PRYES Agar Dehydrated Culture Media, Himedia
Supplier: HiMedia Laboratories
M1172-500G M1172-500G
95020-628EA 282.1 USD InStock
CA95020-628 95020-628
PRYES Agar Dehydrated Culture Media, Himedia
Media for Microbiology Dehydrated Media for Microbiology
For the cultivation and differentiation of nephrotoxin producing strains of Penicillium viridicatum and related species isolated from foods in accordance with APHA.
  • Microbiological culture media
  • 500 g poly bottle
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty