Would you like to log in again?
PA Broth Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia
M1186-500G M1186-500G
95020-652EA 214.03 USD InStock
CA95020-652 95020-652
PA Broth Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For detection of the presence and absence of coliform bacteria in water from treatment plants or distribution systems.
  • Microbiological culture media
  • 500 g poly bottles
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty