Would you like to log in again?
Lysine Arginine Iron Agar Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia Laboratories
M1230-500G M1230-500G
95020-748EA 446.7 USD InStock
CA95020-748 95020-748
Lysine Arginine Iron Agar Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For the isolation and presumptive identification of Yersinia species from milk and milk products.
  • Microbiological culture media
  • 500 g poly bottles
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty