Would you like to log in again?
Duncan Strong (DS) Medium, Modified, Dehydrated Culture Media, HiMedia
Supplier: HiMedia
M1237-500G M1237-500G
95020-762EA 515.17 USD InStock
CA95020-762 95020-762
Duncan Strong (DS) Medium, Modified, Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
For the isolation and differentiation of Clostridium perfringens from other clostridia in foods based on raffinose fermentation.
  • Microbiological culture media
  • 500 g poly bottles
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty