Would you like to log in again?
Colonization Medium with Dulbecco’s Phosphate Buffer, Dehydrated Culture Media, HiMedia
Supplier: HiMedia Laboratories
M1239-500G M1239-500G
95020-766EA 278.05 USD InStock
CA95020-766 95020-766
Colonization Medium with Dulbecco’s Phosphate Buffer, Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
For the preparation of suspension of enterotoxigenic Escherichia coli used for HeLa cell test for colonization.
  • Microbiological culture media
  • 500 g poly bottles
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty