Would you like to log in again?
EC Broth Dehydrated Culture Media, HiMedia
Supplier: HiMedia
M127-500G GM127-5KG M127I-500G GM127-500G M127I-500G M127-500G GM127-2.5KG
95020-836EA 119.95 USD InStock
CA95020-836 95020-836 CA95020-856 75804-082 75804-084 75804-086 95020-856
EC Broth Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
EC Broth is recommended for the selective enumeration of presumptive E. coli by MPN technique from water samples and from clinical samples.
  • Microbiological culture media
Certifications: Complies as per ISO 17025:2005.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty