Would you like to log in again?
Pantothenate Assay Medium, AOAC, Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia Laboratories
M1281-500G M1281-500G
95020-858EA 629.16 USD InStock
CA95020-858 95020-858
Pantothenate Assay Medium, AOAC, Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For microbiological assay of pantothenic acid or salts using Lactobacillus plantarum ATCC 8014 as the test organism.
  • Microbiological culture media
  • 500 g poly bottles
Recommended by AOAC.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty