Would you like to log in again?
M-Aeromonas Selective Agar Base, Dehydrated Culture Media, HiMedia
Supplier: HiMedia
M1283-500G M1283-500G
95020-862EA 257.24 USD InStock
CA95020-862 95020-862
M-Aeromonas Selective Agar Base, Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
For the detection of Aeromonas species in water and other liquid samples by membrane filter technique.
  • Microbiological culture media
  • 500 g poly bottles
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty