Would you like to log in again?
APRY Agar Base Dehydrated Culture Media, HiMedia
Supplier: HiMedia Laboratories
M1291-500G M1291-500G
95020-882EA 256.42 USD InStock
95020-882 CA95020-882
APRY Agar Base Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
For the detection and isolation of acid resistant yeasts, Zygosaccharomyces bailii, and Zygosaccharomyces rouxii in salads, sauces, and dressings
  • Microbiological culture media
  • Packaged in 500 g poly bottles
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty