Would you like to log in again?
Clostridium Broth Base Dehydrated Culture Media, HiMedia
Supplier: HiMedia Laboratories
M1315-500G M1315-500G
95020-918EA 235.23 USD InStock
CA95020-918 95020-918
Clostridium Broth Base Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
For the identification of spores of Clostridium tyrobutyricum which is usually responsible for late blowing in cheese.
  • Microbiological culture media
  • 500 g poly bottles
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty