Would you like to log in again?
Micro Vitamin Test Culture Agar, Dehydrated, HiMedia Laboratories
Supplier: HiMedia Laboratories
M132-500G M132-500G
95020-928EA 188.41 USD InStock
CA95020-928 95020-928
Micro Vitamin Test Culture Agar, Dehydrated, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For the cultivation and maintenance of stock cultures of lactobacilli used in microbiological assay of vitamins.
  • Microbiological culture media
  • 500 g poly bottles
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty