Would you like to log in again?
Shigella Broth Base, HiMedia
Supplier: HiMedia Laboratories
M1326-500G M1326-500G
95020-940EA 317.07 USD InStock
95020-940 CA95020-940
Shigella Broth Base, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
Shigella Broth Base is used for the isolation and cultivation of Shigella species from food.
  • Contains casein enzymic hydrolysate as a source of carbon, nitrogen, vitamins and minerals
  • Buffering provided by dipotassium hydrogen phosphate and potassium dihydrogen phosphate
  • Sodium chloride maintains the osmotic balance of the medium
Shigella are gram-negative, non-motile, non-spore forming rod-shaped bacteria closely related to Escherichia coli and Salmonella. Shigella infection is typically via ingestion (faecal-oral contamination), depending on age and condition of the host, as few as ten bacterial cells can be enough to cause an infection. Shigella causes dysentery that results in the destruction of the epithelial cells of the intestinal mucosa in the cecum and rectum. Some strains produce enterotoxin and Shiga toxin, similar to the verotoxin of E. coli O157:H7. Shigella Broth Base is used for the isolation and cultivation of Shigella species, as recommended by APHA. Shigella Broth Base contains casein enzymic hydrolysate as a source of carbon, nitrogen, vitamins and minerals. Dextrose provides the necessary carbohydrates. Buffering action in the medium is provided by dipotassium hydrogen phosphate and potassium dihydrogen phosphate. Sodium chloride maintains the osmotic balance of the medium. Polysorbate 80 is inhibitory for growth of accompanying microflora besides providing growth factors. Novobiocin is inhibitory for gram-positive bacteria such as S. aureus and certain gram-negative organisms such as H. influenzae and some species of Proteus.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty