Would you like to log in again?
Phosphate Buffer, Modified, Dehydrated Culture Media, Himedia
Supplier: HiMedia Laboratories
M1341-500G M1341-500G
95020-970EA 199.42 USD InStock
95020-970 CA95020-970
Phosphate Buffer, Modified, Dehydrated Culture Media, Himedia
Media for Microbiology Dehydrated Media for Microbiology
A washing buffer solution required during detection of Escherichia coli O157:H7 from food and animal foodstuffs.
  • Microbiological culture media
  • 500 g poly bottle
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty