Would you like to log in again?
MUD SF Broth Base Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia Laboratories
M1343-500G M1343-500G
95020-974EA 413.65 USD InStock
95020-974 CA95020-974
MUD SF Broth Base Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For the detection and enumeration of intestinal enterococci in surface and wastewater by miniaturized method (MPN).
  • Microbiological culture media
  • 500 g poly bottles
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty