Would you like to log in again?
Antibiotic Assay Medium E Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia Laboratories
MM006-500G M1347-500G M1347B-500G M1347-500G MAP006-500G MAP1347-500G MAP1347-500G ME1347-500G
95022-650EA 262.1 USD InStock
CA95022-650 95022-650 95020-984 CA95022-680 95022-680 CA95020-986 CA95020-984 95020-986
Antibiotic Assay Medium E Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Anaerobic basal broth is used for the growth of anaerobic microorganisms, particularly Bacteroides species and other fastidious anaerobes.
  • Microbiological culture media
  • Packaged in 500 g poly bottles
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty