Would you like to log in again?
Aseptic Packing Line Medium Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia Laboratories
M1350-500G M1350-500G
95020-992EA 231.32 USD InStock
95020-992 CA95020-992
Aseptic Packing Line Medium Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
A fluid medium used in validating aseptic packing lines
  • Microbiological culture media
  • Packaged in 500 g poly bottles
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty