Would you like to log in again?
Pantothenate Culture Agar, Dehydrated, HiMedia Laboratories
Supplier: HiMedia Laboratories
M135-500G M135-500G
95020-994EA 279.35 USD InStock
CA95020-994 95020-994
Pantothenate Culture Agar, Dehydrated, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For culturing Lactobacillus plantarum ATCC 8014 used in the microbiological assay of pantothenic acid or salts.
  • Microbiological culture media
  • 500 g poly bottles
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty