Would you like to log in again?
ECD Agar Dehydrated Culture Media, HiMedia
Supplier: HiMedia
M1357-500G M1357-500G
95021-008EA 236.16 USD InStock
CA95021-008 95021-008
ECD Agar Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
For the selective detection of coliforms, especially Escherichia coli in water, food, and other materials and in membrane filter technique.
  • Microbiological culture media
  • 500 g poly bottles
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty