Would you like to log in again?
Water Agar, HiMedia Laboratories
Supplier: HiMedia Laboratories
M1366-2.5KG M1366-2.5KG M1366-500G M1366-5KG M1366-5KG
89223-868EA 551.15 USD InStock
CA89223-870 89223-868 CA89223-868 95021-024 89223-870
Water Agar, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Water Agar is recommended for enumeration, cultivation and observation of sporulation of some fungi.
  • Appearance
  • : White to cream homogeneous free flowing powder
  • Gelling: Firm, comparable with 2.0% agar gel
  • Colour and clarity of prepared medium: White coloured clear to slightly opalescent gel forms in petri plates
Water Agar is often recommended for enumeration, cultivation and observation of sporulation of some fungi
Water Agar is used for enumeration of fungi according to MPN method.
Direct Plating is considered to be one of the most effective techniques for mycological examination of all foods.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty