Would you like to log in again?
Malt Extract Agar Base, Dehydrated Culture Media, HiMedia
Supplier: HiMedia Laboratories
M137-500G M137-500G
95021-042EA 221.38 USD InStock
95021-042 CA95021-042
Malt Extract Agar Base, Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
For the detection, isolation, and enumeration of yeasts and molds.
  • Microbiological culture media
  • 500 g poly bottles
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty