Would you like to log in again?
MUG MFC Agar Dehydrated Culture Media, HiMedia
Supplier: HiMedia Laboratories
M1387-500G M1387-500G
95021-068EA 413.65 USD InStock
95021-068 CA95021-068
MUG MFC Agar Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
For cultivating and enumerating fecal coliforms by the membrane filter technique at elevated temperatures.
  • Microbiological culture media
  • 500 g poly bottles
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty