Would you like to log in again?
Yeast Nitrogen Base, with Amino Acids, HiMedia Laboratories
Supplier: HiMedia Laboratories
M139-500G M139-500G M139-100G M139-100G
95021-078EA 652.08 USD InStock
95021-078 CA95021-078 CA61000-022 61000-022
Yeast Nitrogen Base, with Amino Acids, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Yeast Nitrogen Bases are recommended for classification of yeasts on the basis of their ability to assimilate carbon compounds.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty