Would you like to log in again?
Kohn Two Tube Medium No. 1, Base, Dehydrated, HiMedia Laboratories
Supplier: HiMedia Laboratories
M142-500G M142-500G
95021-118EA 382.97 USD InStock
CA95021-118 95021-118
Kohn Two Tube Medium No. 1, Base, Dehydrated, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For the identification of Enterobacteriaceae based on dextrose and mannitol fermentation and urease production.
  • Microbiological culture media
  • 500 g poly bottles
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty