Would you like to log in again?
Hottinger Broth Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia
M1425-500G M1425-500G
95021-124EA 754.23 USD InStock
95021-124 CA95021-124
Hottinger Broth Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For the cultivation of less fastidious microorganisms and determination of indole in accordance with USSR State Pharmacopoeia.
  • Microbiological culture media
  • 500 g poly bottles
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty