Would you like to log in again?
Percoll™ PLUS Centrifugation Media
Supplier: Cytiva
Percoll™ PLUS
17-5445-01 17-5445-02 17-5445-02 17-5445-01
95021-198EA 348.45 USD InStock
CA95021-200L 95021-198 CA95021-198L 95021-200
Percoll™ PLUS Centrifugation Media
Media for Microbiology Prepared Media for Microbiology
Percoll™ PLUS centrifugation media feature gradients that can either be preformed or spontaneously generated by centrifugation at moderate speeds in an angle-head rotor. Gradients can be made iso-osmotic throughout and cover a range of densities up to 1.3 g/ml. Low viscosity allows cell isolations on preformed gradients in only a few minutes using low centrifugal forces (200 to 1000 g). Media are nontoxic to cells and adjustable for ionic stregth and pH. Intended for in vitro research use only.
Percoll™ medium is used for separation of cells, subcellular particles, and larger viruses (down to ~70S) under gentle conditions that preserve viability and morphological integrity. Percoll™ PLUS medium offers maximum flexibility in clinical research. It is used for preparation of cells, subcellular particles and larger viruses. It has verified low endotoxin level with a maximum of 2 EU/ml.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty