Would you like to log in again?
Eugonic LT 100 Broth Base without Tween 80, Dehydrated, HiMedia Laboratories
Supplier: HiMedia Laboratories
M1517-500G M1517-500G
95021-240EA 297.99 USD InStock
95021-240 CA95021-240
Eugonic LT 100 Broth Base without Tween 80, Dehydrated, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For the cultivation of fastidious microorganisms such as Haemophilus, Neisseria, Pasteurella, Brucella, and Lactobacillus species.
  • Microbiological culture media
  • 500 g poly bottles
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty