Would you like to log in again?
Mannitol Selenite Broth with Brilliant Green, Twin Pack, Dehydrated Culture Media, HiMedia
Supplier: HiMedia Laboratories
M1537-500G M1537-500G
95021-260EA 275.28 USD InStock
95021-260 CA95021-260
Mannitol Selenite Broth with Brilliant Green, Twin Pack, Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
For the enrichment of salmonellae from feces, foodstuffs, and other materials.
  • Microbiological culture media
  • 500 g poly bottles
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty