Would you like to log in again?
Lowenstein Jensen Medium Base without Starch, Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia
M1542-500G M1542-500G
95021-266EA 216.01 USD InStock
95021-266 CA95021-266
Lowenstein Jensen Medium Base without Starch, Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Lowenstein Jensen Medium w/o Starch is recommended for susceptibility testing of Mycobacteria with addition of antitubercular drugs. It is recommended for resistance testing by WHO. Lowenstein-Jensen (L-J) Medium without potato starch with drugs incorporated before inspissation is the modification of the International Union Against Tuberculosis (IUAT). Malachite green serves as an inhibitor and also as a pH indicator.Typical Mycobacterium tuberculosis appears granular, rough, warty and dry friable colonies.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty