Would you like to log in again?
Arabinose Agar Base Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia Laboratories
M1576-500G M1576-500G
95021-288EA 249.03 USD InStock
95021-288 CA95021-288
Arabinose Agar Base Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Recommended to differentiate <italic>Enterococcus faecium </italic>from <italic>Enterococcus faecalis
  • Microbiological culture media
  • Packaged in 500 g poly bottles
</italic>
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty