Would you like to log in again?
Cystine Tryptone Agar Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia Laboratories
M159-500G M159-500G
95021-312EA 132.65 USD InStock
95021-312 CA95021-312
Cystine Tryptone Agar Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For maintenance, subculturing, detection of motility, and fermentation studies with the addition of various carbohydrates.
  • Microbiological culture media
  • 500 g poly bottles
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty