Would you like to log in again?
MRSA Agar Base Dehydrated Culture Media, HiMedia
Supplier: HiMedia Laboratories
M1594-500G M1594-500G
95021-318EA 432.31 USD InStock
95021-318 CA95021-318
MRSA Agar Base Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
For the selection, isolation, and identification of Methicillin Resistant Staphylococcus aureus from clinical specimens.
  • Microbiological culture media
  • 500 g poly bottles
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty