Would you like to log in again?
Costein’s LDS Test Medium, Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia Laboratories
M1621-500G M1621-500G
95021-364EA 272.81 USD InStock
95021-364 CA95021-364
Costein’s LDS Test Medium, Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For the identification of members of Enterobacteriaceae on the basis of lysine decarboxylase and hydrogen sulfide production.
  • Microbiological culture media
  • 500 g poly bottles
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty