Would you like to log in again?
Antimicrobial Inhibitor Test Agar Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia Laboratories
M1601-500G M1631-500G M1632-500G M1632-500G M1601-500G M1631-500G
95021-380EA 292.2 USD InStock
95021-380 95021-382 CA95021-380 CA95021-382 95021-324 CA95021-324
Antimicrobial Inhibitor Test Agar Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For the residual analysis of antimicrobial components in meat and organ samples using Bacillus subtilis (ATCC 6633) and Micrococcus luteus (ATCC 9341) as test organisms
  • Microbiological culture media
  • Packaged in 500 g poly bottles
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty