Would you like to log in again?
Milk Agar Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia Laboratories
M163-500G M163-500G
95021-384EA 256.98 USD InStock
95021-384 CA95021-384
Milk Agar Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For the enumeration of bacteria in milk and milk products, rinse waters, ice cream, and similar products.
  • Microbiological culture media
  • 500 g poly bottles
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty