Would you like to log in again?
Anaerobic Basal Agar Dehydrated Culture Media, HiMedia
Supplier: HiMedia Laboratories
M1635-5KG M1635-500G M1635-5KG M1635-500G M1635-2.5KG M1635-2.5KG
89224-484EA 2180.75 USD InStock
89224-484 CA89224-482 89224-482 95021-386 CA95021-386 CA89224-484
Anaerobic Basal Agar Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
For the growth of anaerobic microorganisms, particularly Bacteroides species and other fastidious anaerobes.
  • Microbiological culture media
Packaging: Packaged in 500 g poly bottles.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty