Would you like to log in again?
Anaerobic Basal Broth Dehydrated Culture Media, HiMedia
Supplier: HiMedia Laboratories
M1636-5KG M1636-2.5KG M1636-2.5KG M1636-500G M1636-5KG M1636-500G
89224-490EA 2002.88 USD InStock
CA89224-490 89224-490 95021-388 CA89224-488 CA95021-388 89224-488
Anaerobic Basal Broth Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
Anaerobic basal broth is used for the growth of anaerobic microorganisms, particularly Bacteroides species and other fastidious anaerobes.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty