Would you like to log in again?
Rappaport Vassiliadis Medium for Water Testing, Modified, Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia Laboratories
M1658I-500G M1658I-500G
95021-412EA 340.46 USD InStock
95021-412 CA95021-412
Rappaport Vassiliadis Medium for Water Testing, Modified, Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For the selective enrichment of Salmonella from water, food, and environmental specimens.
  • Microbiological culture media
  • 500 g poly bottle
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty