Would you like to log in again?
TCBS Agar, HiMedia
Supplier: HiMedia Laboratories
M189-500G M189-2.5KG M189-500G M189-5KG GM189-2.5KG M189-5KG M189-2.5KG GM189-500G GM189-5KG
95026-832EA 1392 USD InStock
CA95026-832 95026-832 95021-464 CA89224-732 CA95021-464 75804-232 89224-732 75804-234 75804-236
TCBS Agar, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
TCBS agar, granulated is recommended for the selective isolation and cultivation of V. cholerae and other enteropathogenic Vibrio's causing food poisoning.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty