Would you like to log in again?
Brain Heart Infusion with PABA and Agar, Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia Laboratories
M213-500G M213-500G
95021-498EA 263.63 USD InStock
95021-498 CA95021-498
Brain Heart Infusion with PABA and Agar, Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For the improved growth of pathogens from the blood of patients undergoing sulfonamide treatment.
  • Microbiological culture media
  • Packaged in 500 g poly bottles
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty