Would you like to log in again?
AK #2 Agar Dehydrated Culture Media, HiMedia
Supplier: HiMedia Laboratories
M234-500G M234-500G
95021-534EA 260.08 USD InStock
95021-534 CA95021-534
AK #2 Agar Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
For the production of spores of Bacillus subtilis ATCC 6633, which are used as inoculum in detection of penicillin and other antibiotics residues in milk and dairy products
  • Microbiological culture media
  • Packaged in 500 g poly bottles
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty