Would you like to log in again?
Antibiotic Assay Medium No. 12 Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia Laboratories
M280-500G M280-500G
95021-604EA 256.98 USD InStock
95021-604 CA95021-604
Antibiotic Assay Medium No. 12 Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For the microbiological assay of amphotericin B and nystatin using <italic>Saccharomyces cerevisiae
  • Microbiological culture media
  • Packaged in 500 g poly bottles
</italic>
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty