Would you like to log in again?
EMB Agar Base Dehydrated Culture Media, HiMedia
Supplier: HiMedia Laboratories
M301-500G M301-500G
95021-648EA 237.05 USD InStock
95021-648 CA95021-648
EMB Agar Base Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
Basal medium to which different carbohydrates and other test substances may be added for differentiation and study of various enteric bacteria.
  • Microbiological culture media
  • 500 g poly bottles
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty