Would you like to log in again?
Tetrathionate Broth Base, Hajna, HiMedia Laboratories
Supplier: HiMedia Laboratories
M327-5KG M327-500G M327-2.5KG M327-2.5KG M327-5KG M327-500G
95021-682EA 149.15 USD InStock
CA89135-076 95021-682 89135-076 CA95021-682 89225-178 CA89225-178
Tetrathionate Broth Base, Hajna, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Tetrathionate broth base, Hajna is used for enrichment and isolation of Salmonellae.
  • Reaction of 9.15% w/v aqueous solution at 25 °C, pH 7.6 ±0.2
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty