Would you like to log in again?
Decarboxylase Broth Base, Moeller, Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia Laboratories
M393-500G M393-500G
95021-768EA 213.29 USD InStock
CA95021-768 95021-768
Decarboxylase Broth Base, Moeller, Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Used to differentiate bacteria based on the ability to decarboxylate an amino acid.
  • Microbiological culture media
  • 500 g poly bottles
Requires the addition of appropriate L-amino acid.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty