Would you like to log in again?
Blue Agar, Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia Laboratories
M401-500G M401-500G
95021-776EA 223.51 USD InStock
95021-776 CA95021-776
Blue Agar, Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
A general-purpose nutrient medium containing an acid base indicator
  • Microbiological culture media
  • Packaged in 500 g poly bottles
With appropriate additions, the medium can be used in carbohydrate fermentation studies.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty