Would you like to log in again?
Universal Beer Agar (UB Agar), HiMedia Laboratories
Supplier: HiMedia Laboratories
M415-500G M415-500G
95021-796EA 218.7 USD InStock
95021-796 CA95021-796
Universal Beer Agar (UB Agar), HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For the cultivation of microorganisms significant in the brewing industry.
  • Detect microbial contamination
  • Helps in growth of variants of lactic acid bacteria
  • Supports the growth of Lactobacilli, Pediococci, Acetobacter and Lymomonas species
Kozulis and Page developed Universal Beer Agar Medium, a basal medium to which beer is added
Universal Beer Agar supports the growth of Lactobacilli, Pediococci, Acetobacter and Lymomonas species and wild yeast strains which may be found infecting the pitching yeast, the cooled wort or during fermentation or storage of the finished beer.
Due to the presence of beer in these media, it is selective for growth of microorganisms that have adapted themselves to the existent conditions in the brewery. The presence of hop constituents and alcohol inhibits growth of many airborne microorganisms not adapted to this environment. Yeast extract is a source of trace elements, vitamins and amino acids. Peptonized milk contains lactose as an energy source. Tomato juice is a source of carbon, protein and nutrients. Dextrose provides additional carbon. Dipotassium and monopotassium phosphates provide buffering capability. Magnesium sulphate, ferrous sulphate and manganese sulphate are sources of ions that simulate metabolism. Sodium chloride maintains the osmotic equilibrium.
The presence of spoilage microorganisms in pitching yeast may be detected from diluted samples by direct surface plating or by pour plate techniques. Incubate the plates aerobically and anaerobically.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty