Would you like to log in again?
Yeast Malt Broth Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia Laboratories
M425-2.5KG M425-500G M425-5KG M425-2.5KG M425-500G M425-5KG
89225-482EA 638.67 USD InStock
89225-482 CA89225-482 89225-484 95021-806 CA95021-806 CA89225-484
Yeast Malt Broth Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Yeast malt broth (YM broth) is used for the isolation and cultivation of yeasts, moulds and other aciduric microorganisms.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty