Would you like to log in again?
Ethyl Violet Azide Broth Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia Laboratories
M426-500G M426S-500G M426-500G M426S-500G
95021-808EA 163.91 USD InStock
95021-808 CA95021-808 95021-810 CA95021-810
Ethyl Violet Azide Broth Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For the selective confirmatory detection of enterococci as an indicator of fecal pollution in water and other specimens.
  • Microbiological culture media
  • 500 g poly bottles
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty