Would you like to log in again?
Soil Extract Agar, HiMedia Laboratories
Supplier: HiMedia Laboratories
M455-2.5KG M455-5KG M455-2.5KG M455-500G M455-500G M455-5KG
95021-852EA 220.07 USD InStock
CA89225-576 95021-852 89225-574 CA95021-852 CA89225-574 89225-576
Soil Extract Agar, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Soil extract agar is used for isolation of soil microorganisms.
  • Cream to yellow homogeneous free flowing powder
  • Firm gelling, comparable with 1.5% agar gel
  • Cultural characteristics can be observed after an incubation at 30 °C for up to 4 days
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty