Would you like to log in again?
Violet Red Bile Broth, HiMedia Laboratories
Supplier: HiMedia Laboratories
M458-5KG M458-500G M458-2.5KG M458-2.5KG M458-5KG M458-500G
95021-858EA 182.88 USD InStock
95021-858 CA95021-858 89225-588 CA89225-586 CA89225-588 89225-586
Violet Red Bile Broth, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Violet red bile broth is used for the detection and enumeration of coliform organisms from water and food.
  • Light yellow to pinkish beige homogeneous free flowing powder
  • pH of 7.20 - 7.60
Violet red bile broth, a modification of MacConkeys original formulation for the enumeration of coli-aerogens bacterial group. It relies on the use of the selective inhibitory components crystals violet and bile salts and the indicator system lactose, and neutral red. The growth of many unwanted organisms is suppressed, while tentative identification of sought bacteria can be made. Lactose non-fermenters and late lactose fermenters produce pale coloured medium.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty